5-Chlor-1,10-phenanthrolin
Lieferant: Thermo Fisher Scientific
31180.02
31180.04
31180.02EA
80.3
EUR
31180.02
31180.04
5-Chlor-1,10-phenanthrolin
5-Chloro-1,10-phenanthroline
CAS No.: 4199-89-7
Molecular Formula: C12H7N2Cl
Formula Weight: 214.66
Physical Form: Crystalline
Sensitivity: Hygroscopic
Melting Point: 123°
MDL No.: MFCD00004980
Formel:
C₁₂H₇ClN₂ Schmelzpunkt: 123 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00004980 CAS-Nummer: 4199-89-7 EINECS: 224-098-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...