L(-)-Tyrosin 99+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH
140640010
140640050
140641000
140640025
ACRO140640010EA
446
EUR
ACRO140640010
ACRO140640050
ACRO140641000
ACRO140640025
L(-)-Tyrosin 99+%
L(-)-Tyrosin
Formel:
C₉H₁₁NO₃ Molekulargewicht: 181.19 g/mol Siedepunkt: 351 Schmelzpunkt: 290 °C (Zers.) Flammpunkt: 176 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00002606 CAS-Nummer: 60-18-4 EINECS: 200-460-4 Merck Index: 13,09907 |
Spezifikation Testergebnisse
Appearance | White to almost white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % (1 g, 105°C) |
Sulfated ash | ≤0.1 % |
Specific optical rotation | -9.8° to -12.3° (20°C, 589 nm) (c=5, 1 N HCl) |
Titration with HClO4 | ≥99.0 % (on dried substance) |
Chloride (Cl) | ≤400 ppm |
Sulfate (SO4) | ≤300 ppm |
Ammonium (NH4) | ≤200 ppm |
Iron (Fe) | ≤10 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...