D(+)-Tryptophan 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
H-D-Trp-OH
168630050
168630250
168631000
ACRO168630050EA
44
EUR
ACRO168630050
ACRO168630250
ACRO168631000
D(+)-Tryptophan 99%
D(+)-Tryptophan
Formel:
C₁₁H₁₂N₂O₂ Molekulargewicht: 204.23 g/mol Schmelzpunkt: 282…285 °C (Zers.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00005647 CAS-Nummer: 153-94-6 EINECS: 205-819-9 Merck Index: 14,09797 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % |
Solubility | (1 % in water) Clear colorless to light yellow |
Specific optical rotation | +29.5° to +33.5° (20°C, 589 nm) (c=1, H2O) |
Titration with HClO4 | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...