(S)-(+)-3-Methyl-2-butanol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (S)-3-Methylbutan-2-ol

L09431.MC L09431.ME
L09431.MCEA 47.8 EUR
L09431.MC L09431.ME
(S)-(+)-3-Methyl-2-butanol ≥99%
(S)-(+)-3-Methyl-2-butanol

Purity: 99% CAS No.: 1517-66-4 Molecular Formula: C5H12O Formula Weight: 88.15 Flash Point: 26°(78°F) Refractive Index: 1.4090 Boiling Point: 112° Beilstein Reference: 1718801 Optical Rotation: +5° (neat) UN No.: 1105 MDL No.: MFCD00064271

Formel: C₅H₁₂O
Siedepunkt: 112 °C
Dichte: 0.810
Flammpunkt: 26 °C (78 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00064271
CAS-Nummer: 1517-66-4
UN-Nummer: 1105
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR