L(+)-Isoleucin

Lieferant: Roth Carl

Synonyme: L-Isoleucine, (2S,3S)-(+)-Isoleucine, H-Ile-OH

3922.1 3922.2 3922.3
ROTH3922.1EA 0 EUR
ROTH3922.1 ROTH3922.2 ROTH3922.3
L(+)-Isoleucin
L(+)-Isoleucin
Formel: H₃CCH₂CH(CH₃)CH(NH₂)COOH
Schmelzpunkt: 279…280 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00064222
CAS-Nummer: 73-32-5
EINECS: 200-798-2
Merck Index: 14,05179

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR