L(+)-Isoleucin 99%

Lieferant: Thermo Fisher Scientific

Synonyme: L-Isoleucine, (2S,3S)-(+)-Isoleucine, H-Ile-OH

166170250 166171000 166170050 166175000
ACRO166170250EA 69.6 EUR
ACRO166170250 ACRO166171000 ACRO166170050 ACRO166175000
L(+)-Isoleucin 99%
L(+)-Isoleucin
Formel: H₃CCH₂CH(CH₃)CH(NH₂)COOH
Molekulargewicht: 131.17 g/mol
Schmelzpunkt: 284…286 °C (Zers.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00064222
CAS-Nummer: 73-32-5
EINECS: 200-798-2
Merck Index: 14,05179

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.3 % (105°C, 3 hrs)
Residue after ignition ≤0.3 %
Specific optical rotation +38.9° to +41.8° (20°C, 589 nm) (c=4, 6 N HCl)
Titration with HClO4 98.5 to 101.5 %
Chloride (Cl) ≤500 ppm
Sulfate (SO4) ≤300 ppm
Iron (Fe) ≤30 ppm
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR