L(+)-allo-Isoleucin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(2S,3R)-(+)-allo-Isoleucine, L-Alloisoleucine, (2S,3R)-2-Amino-3-methylpentanoic acid
M-Clarity™ quality level = MQ200
I8754-100MG
I8754-1G
I8754-25MG
SIALI8754-100MGEA
250
EUR
SIALI8754-100MG
SIALI8754-1G
SIALI8754-25MG
L(+)-allo-Isoleucin, Sigma-Aldrich®
L(+)-allo-Isoleucin
Formel:
H₃CCH₂CH(CH₃)CH(NH₂)COOH |
MDL Number:
MFCD00066446 CAS-Nummer: 1509-34-8 EINECS: 216-142-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...