D(-)-allo-Isoleucin 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
H-D-Allo-Ile-OH, (2R,3S)-(-)-allo-Isoleucine, H-D-allo-Ile-OH
357430010
357435000
ACRO357430010EA
239
EUR
ACRO357430010
ACRO357435000
D(-)-allo-Isoleucin 97%
D(-)-allo-Isoleucin
Formel:
C₆H₁₃NO₂ Schmelzpunkt: 285 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00066445 CAS-Nummer: 1509-35-9 EINECS: 216-143-9 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Specific optical rotation | -35° to -39° (20°C, 589 nm) (C=5, 1 N HCl) |
Titration with HClO4 | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...