D(-)-Mannitol ACS
Lieferant: Thermo Fisher Scientific
Synonyme:
Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
CAS No.: 69-65-8
Molecular Formula: HOCH2(CHOH)4CH2OH
Formula Weight: 182.17
Physical Form: Crystalline
Density: 1.489
Appearance: White
Odour: Odourless
Melting Point: 165-167°
Boiling Point: 290-295°/3.5mm
Solubility: Soluble in water, alcohol, pyridine, aniline. Insoluble in ether
Beilstein Reference: 1721898
Merck Reference: 14,5745
MDL No.: MFCD00064287
Formel:
HOH₂C(CH(OH))₄CH₂OH Molekulargewicht: 182.17 g/mol Siedepunkt: 290…295 °C (3.5 mmHg) Schmelzpunkt: 165…167 °C Dichte: 1.489 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00064287 CAS-Nummer: 69-65-8 EINECS: 200-711-8 Merck Index: 14,5745 |