(S)-(+)-2-Butanol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (S)-(+)-sec-Butyl alcohol, (S)-Butan-2-ol, (S)-(+)-sec-Butanol, (S)-(+)-Ethyl methyl carbinol, (S)-(+)-2-Hydroxy butane, (S)-(+)-Methyl ethyl carbinol

L14164.03 L14164.06
L14164.03EA 71.1 EUR
L14164.03 L14164.06
(S)-(+)-2-Butanol ≥98%
(S)-(+)-2-Butanol

Purity: 98+% CAS No.: 4221-99-2 Molecular Formula: C4H10O Formula Weight: 74.12 Flash Point: 26°(79°F) Refractive Index: 1.3980 Odour: Alcohol-like Boiling Point: 99-100° Beilstein Reference: 1718763 Merck Reference: 14,1541 Optical Rotation: +13.5° (c=10 in methanol) UN No.: 1120 MDL No.: MFCD00064281

Formel: C₄H₁₀O
Siedepunkt: 99…100 °C
Schmelzpunkt: –114 °C
Dichte: 0.806
Flammpunkt: 26 °C (79 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00064281
CAS-Nummer: 4221-99-2
EINECS: 224-168-1
UN-Nummer: 1120
ADR: 3,III
Merck Index: 14,1541

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR