di-Ammonium-L(+)-tartrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(R,R)-(+)-Diammonium tartrate, (R,R)-(+)-Ammonium tartrate dibasic, Ammonium tartrate dibasic, L(+)-Tartaric acid diammonium salt, (+)-Diammonium L-tartrate, (R,R)-(+)-Tartaric acid diammonium salt, L(+)-Ammonium tartrate dibasic, Ammonium L-(+)-tartrate
M-Clarity™ quality level = MQ200
09985-1KG
09985-250G
09985-50G
SIAL09985-1KGEA
327
EUR
SIAL09985-1KG
SIAL09985-250G
SIAL09985-50G
di-Ammonium-L(+)-tartrat, Sigma-Aldrich®
di-Ammonium-L(+)-tartrat
Formel:
C₄H₁₂N₂O₆ Dichte: ∼1.601 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00013073 CAS-Nummer: 3164-29-2 EINECS: 221-618-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...