Kalium-D-gluconat ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Potassium Gluconate, Gluconic Acid Potassium Salt, D-Gluconic acid potassium salt, Potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
B25135.0B
B25135.18
B25135.30
B25135.0BEA
130
EUR
B25135.0B
B25135.18
B25135.30
Kalium-D-gluconat ≥99%
Kalium-D-gluconat
Purity: 99%
CAS No.: 299-27-4
Molecular Formula: C6H11KO7
Formula Weight: 234.25
Melting Point: ca 180° dec.
MDL No.: MFCD00064211
Formel:
HOH₂C(CH(OH))₄COOK Schmelzpunkt: ∼ 180 °C (dec.) Dichte: 1.73 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00064211 CAS-Nummer: 299-27-4 EINECS: 206-074-2 Merck Index: 12,07796 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...