D(-)-Norleucin ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: H-D-Nle-OH, (R)-(-)-Norleucine

L08257.03 L08257.06
L08257.03EA 48.1 EUR
L08257.03 L08257.06
D(-)-Norleucin ≥99%
D(-)-Norleucin

Purity: 99% CAS No.: 327-56-0 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: 300° Beilstein Reference: 1721749 Merck Reference: 14,6706 Optical Rotation: -23° (c=5 in 5N HCl) MDL No.: MFCD00008099

Formel: C₆H₁₃NO₂
Molekulargewicht: 131.17 g/mol
Schmelzpunkt: >300 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00008099
CAS-Nummer: 327-56-0
EINECS: 206-320-9
Merck Index: 12,06800

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR