L(+)-Arginin ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH
A15738.0E A15738.14 A15738.22 A15738.36 AB110500.0100
A15738.0EEA 625 EUR
A15738.0E A15738.14 A15738.22 A15738.36 ABCRAB110500.0100
L(+)-Arginin ≥98%
L(+)-Arginin
Purity: 98+% CAS No.: 74-79-3 Molecular Formula: C6H14N4O2 Formula Weight: 174.20 Physical Form: Powder Sensitivity: Air Sensitive Appearance: White Melting Point: 226° dec. Beilstein Reference: 1725413 Merck Reference: 14,780 Optical Rotation: +26° (c=1.6 in 6N HCl) MDL No.: MFCD00002635
Formel: C₆H₁₄N₄O₂
Molekulargewicht: 174.2 g/mol
Siedepunkt: 368
Schmelzpunkt: 226 °C (Zers.)
Dichte: 1.3 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00002635
CAS-Nummer: 74-79-3
EINECS: 200-811-1
Merck Index: 13,00785