L(+)-Diethyltartrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (+)-Diethyl L-tartrate, (R,R)-(+)-Tartaric acid diethyl ester, (2R,3R)-(+)-Diethyl tartrate, Diethyl L-tartrate, L(+)-Tartaric acid diethyl ester, (2R,3R)-Diethyl 2,3-dihydroxysuccinate, (R,R)-(+)-Diethyl tartrate

M-Clarity™ quality level = MQ200

156841-100G 156841-25G 156841-500G
SIAL156841-100GEA 71.2 EUR
SIAL156841-100G SIAL156841-25G SIAL156841-500G
L(+)-Diethyltartrat, Sigma-Aldrich®
L(+)-Diethyltartrat
Formel: C₈H₁₄O₆
Siedepunkt: 280 °C (1013 hPa)
Schmelzpunkt: 18.7 °C
Dichte: 1.21 g/cm³ (20 °C)
MDL Number: MFCD00009143
CAS-Nummer: 87-91-2
EINECS: 201-783-3
Merck Index: 13,03886

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR