(R)-(-)-Linalool, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (-)-Linalool, (R)-3,7-Dimethylocta-1,6-dien-3-ol, (R)-(-)-3,7-Dimethyl-1,6-octadien-3-ol, L-Linalool

M-Clarity™ Qualitätsstufe = MQ300

W263516-1KG-K W263516-4KG-K W263516-9KG-K
SIALW263516-1KG-KEA 155 EUR
SIALW263516-1KG-K SIALW263516-4KG-K SIALW263516-9KG-K
(R)-(-)-Linalool, Sigma-Aldrich®
(R)-(-)-Linalool
Formel: C₁₀H₁₈O
Molekulargewicht: 154.25 g/mol
MDL Number: MFCD00135469
CAS-Nummer: 126-91-0
EINECS: 204-811-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR