Nickel(II)fluorid Tetrahydrat ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Nickel difluoride tetrahydrate

14132.18 14132.30
14132.18EA 60.1 EUR
14132.18 14132.30
Nickel(II)fluorid Tetrahydrat ≥98%
Nickel(II)fluorid Tetrahydrat

Purity: 98+% CAS No.: 13940-83-5 Molecular Formula: NiF2.4H2O Formula Weight: 168.77 (96.71anhy) Physical Form: Powder Odour: Odourless UN No.: 3288 MDL No.: MFCD00016263

Formel: F₂Ni·4H₂O
Molekulargewicht: 168.75 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00016263
CAS-Nummer: 13940-83-5
EINECS: 233-071-3
UN-Nummer: 3288
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR