Rhodium(III)chlorid Hydrat (38,5 - 45,5% Rh)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Rhodium trichloride hydrate

11032.03 11032.06 11032.14 11032.01
11032.03EA 1400 EUR
11032.03 11032.06 11032.14 11032.01
Rhodium(III)chlorid Hydrat (38,5 - 45,5% Rh)
Rhodium(III)chlorid Hydrat

CAS No.: 20765-98-4 Molecular Formula: RhCl3.xH2O Formula Weight: 209.26(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Dark Red Melting Point: 100° dec. Solubility: Soluble in alcohol, water. Slightly soluble in acetone Merck Reference: 14,8188 MDL No.: MFCD00149839

Formel: Cl₃Rh·H₂O
Schmelzpunkt: 100 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00149839
CAS-Nummer: 20765-98-4
EINECS: 233-165-4
Merck Index: 14,8188

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR