Nickel(II)chlorid Hexahydrat ≥99,9995% (Metall-Basis), Puratronic®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Nickel dichloride hexahydrate
43185.22
43185.14
43185.22EA
725
EUR
43185.22
43185.14
Nickel(II)chlorid Hexahydrat ≥99,9995% (Metall-Basis), Puratronic®
Nickel(II)chlorid Hexahydrat
Purity: 99.9995%
CAS No.: 7791-20-0
Molecular Formula: Cl2Ni.6H2O
Formula Weight: 237.71 (129.60anhy)
Physical Form: Crystalline
Density: 1.92
Appearance: Green
Odour: Odourless
pH: 4.9
Melting Point: 140° -H2O
Merck Reference: 14,6505
UN No.: 3288
MDL No.: MFCD00149809
Formel:
NiCl₂·6H₂O Schmelzpunkt: 140 °C (-H₂O) Dichte: 1.92 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00149809 CAS-Nummer: 7791-20-0 EINECS: 231-743-0 UN-Nummer: 3288 ADR: 6.1,III Merck Index: 14,6505 |