Eisen(II)chlorid Tetrahydrat 98,0-102,0%, AnalaR NORMAPUR® analytisches Reagens
Lieferant: VWR Chemicals
|
Gefahr
|
Synonyme:
Ferrous chloride tetrahydrate, Iron dichloride tetrahydrate
24127.237EA
67.5
EUR
24127.237
24127.460
Eisen(II)chlorid Tetrahydrat 98,0-102,0%, AnalaR NORMAPUR® analytisches Reagens
Eisen(II)chlorid Tetrahydrat
Formel:
FeCl₂·4H₂O Schmelzpunkt: 677 °C Dichte: 1.926 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00149709 CAS-Nummer: 13478-10-9 EINECS: 231-843-4 UN-Nummer: 3260 ADR: 8,III |
Spezifikation Testergebnisse
Assay | 98.0 - 102.0 % |
Not precipitated by NH₄OH (as SO₄) | Max. 0.1 % |
Total N (Nitrogen) | Max. 10 ppm |
SO₄ (Sulphate) | Max. 100 ppm |
As (Arsenic) | Max. 5 ppm |
Pb (Lead) | Max. 10 ppm |
Zn (Zinc) | Max. 100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...