Mangan(II)nitrat Hydrat ≥99,999% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Manganous nitrate hydrate

10806.04 10806.09 10806.18
10806.04EA 28.4 EUR
10806.04 10806.09 10806.18
Mangan(II)nitrat Hydrat ≥99,999% (Metall-Basis), Puratronic®
Mangan(II)nitrat Hydrat

Purity: 99.999% CAS No.: 15710-66-4 Molecular Formula: Mn(NO3)2.xH2O Formula Weight: 178.95(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Appearance: Pink Odour: Odourless Melting Point: ca 26° Merck Reference: 14,5733 UN No.: 2724 MDL No.: MFCD00149788

Formel: Mn(NO₃)₂·H₂O
Molekulargewicht: 196.96 g/mol
Schmelzpunkt: ∼ 26 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00149788
CAS-Nummer: 15710-66-4
EINECS: 233-828-8
UN-Nummer: 2724
ADR: 5.1 III
Merck Index: 14,5733

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR