Mangan(II)chlorid Tetrahydrat ≥99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate
11563.22 11563.36
11563.22EA 36.8 EUR
11563.22 11563.36
Mangan(II)chlorid Tetrahydrat ≥99% (Metall-Basis)
Mangan(II)chlorid Tetrahydrat
Purity: 99% CAS No.: 13446-34-9 Molecular Formula: Cl2Mn.4H2O Formula Weight: 197.90 (125.84anhy) Physical Form: Crystalline Density: 2.010 Appearance: Pink Odour: Odourless Melting Point: 58° Merck Reference: 14,5728 MDL No.: MFCD00149792
Formel: MnCl₂·4H₂O
Molekulargewicht: 197.9 g/mol
Siedepunkt: 1190
Schmelzpunkt: 58 °C
Dichte: 2.010
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00149792
CAS-Nummer: 13446-34-9
EINECS: 231-869-6
UN-Nummer: 3077
ADR: 9 III
Merck Index: 14,5728