Mangan(II)bromid Tetrahydrat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Manganous bromide tetrahydrate, Manganese dibromide tetrahydrate, Manganese(II) bromide hydrate
Purity: 98%
CAS No.: 10031-20-6
Molecular Formula: MnBr2.xH2O
Formula Weight: 214.76(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: Pink
Odour: Odourless
Melting Point: 64.3° dec.
Solubility: Soluble in water, alcohol
Merck Reference: 14,5725
MDL No.: MFCD00149791
Formel:
Br₂Mn·4H₂O Molekulargewicht: 286.81 g/mol Schmelzpunkt: 64.3 °C (Zers.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00149791 CAS-Nummer: 10031-20-6 EINECS: 236-591-9 |