Yttrium(III)nitrat Tetrahydrat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Yttrium trinitrate tetrahydrate
M-Clarity™ quality level = MQ100
331309-100G
331309-10G
SIAL331309-100GEA
610
EUR
SIAL331309-100G
SIAL331309-10G
Yttrium(III)nitrat Tetrahydrat, Sigma-Aldrich®
Yttrium(III)nitrat Tetrahydrat
Formel:
N₃O₉Y·4H₂O |
MDL Number:
MFCD00149938 CAS-Nummer: 13773-69-8 EINECS: 233-802-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...