Scandium(III)nitrat-Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Trinitric acid scandium salt hydrate, Scandium(3+) trinitrate hydrate, Scandium trinitrate hydrate

M-Clarity™ quality level = MQ100

325902-1G 325902-5G
SIAL325902-1GEA 233 EUR
SIAL325902-1G SIAL325902-5G
Scandium(III)nitrat-Hydrat, Sigma-Aldrich®
Scandium(III)nitrat-Hydrat
Formel: Sc(NO₃)₃·xH₂O
MDL Number: MFCD00011220
CAS-Nummer: 107552-14-7
EINECS: 236-701-5
UN-Nummer: 1477
ADR: 5.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR