Scandium(III)nitrat-Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Trinitric acid scandium salt hydrate, Scandium(3+) trinitrate hydrate, Scandium trinitrate hydrate
Purity: 99.9%
CAS No.: 13465-60-6
Molecular Formula: Sc(NO3)3.xH2O
Formula Weight: 230.97(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Solubility: Freely soluble in water and alcohol
Merck Reference: 14,8392
UN No.: 1477
MDL No.: MFCD00011220
Formel:
Sc(NO₃)₃·xH₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011220 CAS-Nummer: 107552-14-7 EINECS: 236-701-5 UN-Nummer: 1477 ADR: 5.1,III Merck Index: 14,8392 |