Kupfer(II)fluorid Dihydrat
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Cupric fluoride dihydrate, Copper(II) fluoride hydrate
13076.18
13076.30
13076.18EA
51.7
EUR
13076.18
13076.30
Kupfer(II)fluorid Dihydrat
Kupfer(II)fluorid Dihydrat
CAS No.: 13454-88-1
Molecular Formula: CuF2.2H2O
Formula Weight: 137.57 (101.54anhy)
Physical Form: Powder
Density: 2.934
Odour: Odourless
Merck Reference: 14,2638
UN No.: 3288
MDL No.: MFCD00010977
Flux in casting gray iron
Formel:
CuF₂·2H₂O Molekulargewicht: 137.57 g/mol Schmelzpunkt: >130 °C (Zers.) Dichte: 2.934 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00010977 CAS-Nummer: 13454-88-1 EINECS: 232-147-3 UN-Nummer: 3288 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...