Certrichlorid Heptahydrat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Cerous chloride heptahydrate, Cerium (III) chloride heptahydrate

M-Clarity™ quality level = MQ100

202983-10G 202983-100G
SIAL202983-10GEA 165 EUR
SIAL202983-10G SIAL202983-100G
Certrichlorid Heptahydrat, Sigma-Aldrich®
Certrichlorid Heptahydrat
Formel: CeCl₃·7H₂O
Schmelzpunkt: 848 °C
Dichte: 3.97 g/cm³ (20 °C)
MDL Number: MFCD00149634
CAS-Nummer: 18618-55-8
EINECS: 232-227-8
Merck Index: 12,02041

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR