Ytterbium(III)bromid Hydrat ≥99,9%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
14575.09
14575.18
14575.09EA
109
EUR
14575.09
14575.18
Ytterbium(III)bromid Hydrat ≥99,9%
Ytterbium(III)bromid Hydrat
Purity: 99.9%
CAS No.: 15163-03-8
Molecular Formula: YbBr3.xH2O
Formula Weight: 412.77(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 300°
MDL No.: MFCD00049613
Formel:
Br₃Yb·H₂O Schmelzpunkt: 300 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00049613 CAS-Nummer: 15163-03-8 EINECS: 237-350-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...