Aluminiumfluorid Trihydrat ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Aluminum fluoride trihydrate

11496.22 11496.36 11496.A3
11496.22EA 40.1 EUR
11496.22 11496.36 11496.A3
Aluminiumfluorid Trihydrat ≥97%
Aluminiumfluorid Trihydrat

Purity: 97% CAS No.: 15098-87-0 Molecular Formula: AlF3.3H2O Formula Weight: 138.02 (83.98anhy) Physical Form: Powder Density: 2.17 Appearance: White Odour: Odourless Melting Point: 250° -3H2O Merck Reference: 14,339 MDL No.: MFCD00003426

Formel: AlF₃·3H₂O
Molekulargewicht: 138.02 g/mol
Schmelzpunkt: 250 °C (-3H₂O)
Dichte: 2.17
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00149141
CAS-Nummer: 15098-87-0
EINECS: 232-051-1
UN-Nummer: 3260
ADR: 8 III
Merck Index: 12,00351

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR