Magnesiumperchlorat Hexahydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

309303-100G 309303-500G
SIAL309303-100GEA 37.4 EUR
SIAL309303-100G SIAL309303-500G
Magnesiumperchlorat Hexahydrat, Sigma-Aldrich®
Magnesiumperchlorat Hexahydrat
Formel: Cl₂MgO₈·6H₂O
Schmelzpunkt: 185…190 °C
Dichte: 1.98 g/cm³ (20 °C)
MDL Number: MFCD00149778
CAS-Nummer: 13446-19-0
EINECS: 233-108-3
UN-Nummer: 1475
ADR: 5.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR