Calciumbromid Hydrat ≥95%

Lieferant: Thermo Fisher Scientific
Achtung

18820.06 18820.22 18820.36
18820.06EA 41.5 EUR
18820.06 18820.22 18820.36
Calciumbromid Hydrat ≥95%
Calciumbromid Hydrat

Purity: 95% CAS No.: 71626-99-8 Molecular Formula: CaBr2.xH2O Formula Weight: 199.90(anhy) Physical Form: Powder Appearance: White MDL No.: MFCD00149608

Formel: Br₂Ca·H₂O
Dichte: 2.295 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00149608
CAS-Nummer: 71626-99-8
EINECS: 232-164-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR