Aluminiumkaliumsulfat Dodecahydrat
Lieferant: Roth Carl
Synonyme:
Aluminium Potassium sulphate, 12-Hydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium
8896.1
8896.2
ROTH8896.1EA
0
EUR
ROTH8896.1
ROTH8896.2
Aluminiumkaliumsulfat Dodecahydrat
Aluminiumkaliumsulfat Dodecahydrat
Formel:
AlK(SO₄)₂·12H₂O Schmelzpunkt: 92.5 °C Dichte: 1.72 g/cm³ (20 °C) |
MDL Number:
MFCD00149143 CAS-Nummer: 7784-24-9 EINECS: 233-141-3 Merck Index: 12,00373 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...