Phloroglucin Dihydrat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 1,3,5-Trihydroxybenzene dihydrate, Phloroglucinol, Hydrate

M-Clarity™ quality level = MQ100

P38005-100G P38005-25G
SIALP38005-100GEA 92.5 EUR
SIALP38005-100G SIALP38005-25G
Phloroglucin Dihydrat, Sigma-Aldrich®
Phloroglucin Dihydrat
Formel: C₆H₆O₃·2H₂O
Schmelzpunkt: 218…221 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00149090
CAS-Nummer: 6099-90-7
EINECS: 203-611-2
Merck Index: 13,07413

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR