Phloroglucin Dihydrat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A13034.14
A13034.22
A13034.36
A13034.14EA
29.4
EUR
A13034.14
A13034.22
A13034.36
Phloroglucin Dihydrat ≥98%
Phloroglucin Dihydrat
Purity: 98%
CAS No.: 6099-90-7
Molecular Formula: C6H6O3.2H2O
Formula Weight: 162.14 (126.11anhy)
Odour: Odourless
Melting Point: 214-217°
MDL No.: MFCD00149090
Formel:
C₆H₆O₃·2H₂O Schmelzpunkt: 214…217 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00149090 CAS-Nummer: 6099-90-7 EINECS: 203-611-2 Merck Index: 14,7328 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...