Kupfer(II)acetat Monohydrat, Kristalle, EMPLURA®, Supelco®

Lieferant: Merck
Gefahr

Synonyme: Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate

M-Clarity™ quality level = MQ200

1.02710.0500 1.02710.9050 1.02710.1700
1.02710.0500EA 150 EUR
1.02710.0500 1.02710.9050 1.02710.1700
Kupfer(II)acetat Monohydrat, Kristalle, EMPLURA®, Supelco®
Kupfer(II)acetat Monohydrat
Formel: (H₃CCOO)₂Cu·H₂O
Schmelzpunkt: 115 °C
Dichte: 1.88 g/cm³ (20 °C)
MDL Number: MFCD00149570
CAS-Nummer: 6046-93-1
UN-Nummer: 3077
ADR: 9,III
Merck Index: 13,02651

Bestellen

Spezifikation Testergebnisse

Assay (iodometric) ≥ 99.0 %
In water insoluble matter ≤ 0.1 %
pH-value (2 %; water) 5.2 - 5.5
Chloride (Cl) ≤ 0.01 %
Sulfate (SO₄) ≤ 0.2 %
Fe (Iron) ≤ 0.005 %
Zn (Zinc) ≤ 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR