Kupfer(II)acetat Monohydrat ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate
Purity: 99.9%
CAS No.: 6046-93-1
Molecular Formula: C4H6CuO4.H2O
Formula Weight: 199.65 (181.64anhy)
Physical Form: -4 Mesh
Density: 1.882
Appearance: Blue-green
Odour: Odourless
pH: 5.2-5.5
Melting Point: 115°
Merck Reference: 14,2624
UN No.: 3077
MDL No.: MFCD00149570
Formel:
(H₃CCOO)₂Cu·H₂O Schmelzpunkt: 115 °C Dichte: 1.882 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00149570 CAS-Nummer: 6046-93-1 EINECS: 205-553-3 UN-Nummer: 3077 ADR: 9,III Merck Index: 14,2624 |