Kupfer(II)acetat Monohydrat 98,0-102,0% ACS

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate

14177.22 14177.36 14177.A3
14177.22EA 43.8 EUR
14177.22 14177.36 14177.A3
Kupfer(II)acetat Monohydrat 98,0-102,0% ACS
Kupfer(II)acetat Monohydrat

Purity: 98.0-102.0% CAS No.: 6046-93-1 Molecular Formula: C4H6CuO4.H2O Formula Weight: 199.65 (181.64anhy) Physical Form: Powder Density: 1.882 Appearance: Blue-green Odour: Odourless pH: 5.2-5.5 Melting Point: 115° Merck Reference: 14,2624 UN No.: 3077 MDL No.: MFCD00149570

Formel: (H₃CCOO)₂Cu·H₂O
Schmelzpunkt: 115 °C
Dichte: 1.882
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00149570
CAS-Nummer: 6046-93-1
EINECS: 205-553-3
UN-Nummer: 3077
ADR: 9,III
Merck Index: 14,2624

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR