Natriumacetat Trihydrat 99-101% ACS
Lieferant: Thermo Fisher Scientific
Synonyme:
Acetic acid sodium salt trihydrate
Common Applications: In photography, analytical laboratory as buffer, pharmaceutical
Purity: 99.0%-100.5%
CAS No.: 6131-90-4
Molecular Formula: NaOOCCH3.3H2O
Formula Weight: 136.08 (82.03anhy)
Physical Form: Crystalline
Density: 1.45
pH: 7.5-9.0
Melting Point: 62-64°
Solubility: Freely soluble in alcohol
Beilstein Reference: 3732037
Merck Reference: 14,8571
MDL No.: MFCD00071557
In photography, analytical laboratory as buffer, pharmaceutical
Formel:
H₃CCOONa·3H₂O Siedepunkt: 120…123 °C (1013 hPa) Schmelzpunkt: 62…64 °C Dichte: 1.45 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00071557 CAS-Nummer: 6131-90-4 EINECS: 204-823-8 Merck Index: 14,8571 |