Semicarbazid Hydrochlorid

Lieferant: Roth Carl
Gefahr

Synonyme: Semicarbazide hydrochloride, Carbamylhydrazine hydrochloride, Aminourea hydrochloride, Hydrazinecarboxamide hydrochloride, N-Aminourea hydrochloride, Hydrazinecarboxamidehydrochloride

4681.1 4681.2
ROTH4681.1EA 0 EUR
ROTH4681.1 ROTH4681.2
Semicarbazid Hydrochlorid
Semicarbazid Hydrochlorid
Formel: CH₆ClN₃O
Schmelzpunkt: 174…178 °C
MDL Number: MFCD00013009
CAS-Nummer: 563-41-7
EINECS: 209-247-0
UN-Nummer: 2811
ADR: 6.1,III
Merck Index: 12,08588

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR