Adenin Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
7H-Purin-6-amine hydrochloride, Purin-6-amine hydrochloride, 6-Aminopurine hydrochloride
M-Clarity™ quality level = MQ200
A8751-1G
A8751-100G
A8751-25G
A8751-5G
SIALA8751-1GEA
29.7
EUR
SIALA8751-1G
SIALA8751-100G
SIALA8751-25G
SIALA8751-5G
Adenin Hydrochlorid, Sigma-Aldrich®
Adenin Hydrochlorid
Formel:
C₅H₅N₅·HCl |
MDL Number:
MFCD00150864 CAS-Nummer: 2922-28-3 EINECS: 220-867-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...