Polymyxin B-Sulfat, Pulver USP, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

P0972-10MU P0972-1MU P0972-50MU
SIALP0972-10MUEA 402 EUR
SIALP0972-10MU SIALP0972-1MU SIALP0972-50MU
Polymyxin B-Sulfat, Pulver USP, Sigma-Aldrich®
Polymyxin B sulphate
Formel: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00131991
CAS-Nummer: 1405-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR