Neufuchsin

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Basic Violet 2, New Fuchsin, 4-[(4-Amino-m-tolyl)(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-o-toluidine hydrochloride

212140250 212141000
ACRO212140250EA 38.7 EUR
ACRO212140250 ACRO212141000
Neufuchsin
Neufuchsin
Formel: C₂₂H₂₄ClN₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00012567
CAS-Nummer: 3248-91-7
EINECS: 221-831-7

Bestellen

Spezifikation Testergebnisse

Appearance Dark green Crystalline powder or crystals
Infrared spectrum Conforms
UV Lambda max 550 to 554 nm (in 50% ethanol)
Absorption Ratio P-15/P+15 1.1 to 1.4
Specific absorbance A (1%/1cm) ≥1500 (at lambda max.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR