L(+)-Cystein Hydrochlorid Monohydrat ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(R)-(+)-Cysteine hydrochloride monohydrate, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, L-Cysteine HCI Monohydrate, H-Cys-OH.HCl.H2O, Cysteine Hydrochloride, Monohydrate, Cysteine Hydrochloride monohydrate
Purity: 99%
CAS No.: 7048-04-6
Molecular Formula: C3H7NO2S.HCl.H2O
Formula Weight: 175.64 (157.62anhy)
pH: 1-2
Melting Point: ca 175° dec.
Beilstein Reference: 5158059
Merck Reference: 14,2781
Optical Rotation: +5.5° (c=5 in 5N HCl)
MDL No.: MFCD00065606
Formel:
HSCH₂CH(NH₂)COOH·HCl·H₂O Schmelzpunkt: ∼ 175 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00065606 CAS-Nummer: 7048-04-6 EINECS: 200-157-7 Merck Index: 14,2781 |