Ammoniumcer(IV)nitrat 0.1 mol/l (0.1 N) in wässriger Lösung, Titripur® Reag. Ph Eur volumetrische Lösung, Supelco®
Lieferant: Merck
Gefahr
Synonyme: Ammonium ceric nitrate, Ammonium hexanitratocerate (IV), Ceric Ammonium Nitrate, Cerium (IV) ammonium nitrate, Diammonium hexanitratocerate
M-Clarity™ Qualitätsstufe = MQ100
1.02277.1000 1.02277.1003
1.02277.1000EA 132 EUR
1.02277.1000 1.02277.1003
Ammoniumcer(IV)nitrat 0.1 mol/l (0.1 N) in wässriger Lösung, Titripur® Reag. Ph Eur volumetrische Lösung, Supelco®
Ammoniumcer(IV)nitrat
Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.
Formel: (NH₄)₂Ce(NO₃)₆
Molekulargewicht: 548.22 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00151121
CAS-Nummer: 16774-21-3
EINECS: 240-827-6
UN-Nummer: 2796
ADR: 8 II
Merck Index: 13,00509