Kaliumhydroxid 0.1 mol/l (0.1 N) in Ethanol, Titripur® Reag. USP, Reag. Ph. Eur. volumetrische Lösung, Supelco®
Lieferant: Merck
Gefahr
Synonyme: Potash caustic, Caustic potash
M-Clarity™ Qualitätsstufe = MQ100
1.09115.1000 1.09115.2500 1.09115.1003
1.09115.1000EA 79.5 EUR
1.09115.1000 1.09115.2500 1.09115.1003
Kaliumhydroxid 0.1 mol/l (0.1 N) in Ethanol, Titripur® Reag. USP, Reag. Ph. Eur. volumetrische Lösung, Supelco®
Kaliumhydroxid
Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.
Formel: KOH
Molekulargewicht: 56.11 g/mol
Siedepunkt: 1320
Schmelzpunkt: 360
Dichte: 0.836 g/cm³ (20 °C)
Flammpunkt: 12 °C (EtOH)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003553
CAS-Nummer: 1310-58-3
EINECS: 215-181-3
UN-Nummer: 2924
ADR: 3 (8) II
Merck Index: 12,07806
Bestellen
Spezifikation Testergebnisse
Form liquid
Amount-of-substance concentration 0.0995-0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM