(±)-δ-Decalacton ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: 6-Pentyltetrahydro-2H-pyran-2-one, 5-Decanolide, (±)-Tetrahydro-6-pentyl-2H-pyran-2-one, δ-Decalactone, (±)-Decan-5-olide

A11756.14 A11756.18 A11756.22 A11756.36
A11756.14EA 50.9 EUR
A11756.14 A11756.18 A11756.22 A11756.36
(±)-δ-Decalacton ≥97%
(±)-δ-Decalacton

Purity: 97% CAS No.: 705-86-2 Molecular Formula: C10H18O2 Formula Weight: 170.25 Flash Point: 145°(293°F) Refractive Index: 1.4580 Melting Point: -25° Boiling Point: 117-120°/0.02m Beilstein Reference: 117520 MDL No.: MFCD00006649

Formel: C₁₀H₁₈O₂
Siedepunkt: 117…120 °C (0.02 mmHg)
Schmelzpunkt: –25 °C
Dichte: 0.971
Flammpunkt: 145 °C (293 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00006649
CAS-Nummer: 705-86-2
EINECS: 211-889-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR