5-Hydroxy-DL-tryptophan ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
DL-5-Hydroxytryptophan, 2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
A12237.03
A12237.06
A12237.03EA
67.8
EUR
A12237.03
A12237.06
5-Hydroxy-DL-tryptophan ≥99%
5-Hydroxy-DL-tryptophan
Purity: 99%
CAS No.: 56-69-9
Molecular Formula: C11H12N2O3
Formula Weight: 220.23
Melting Point: ca 298° dec.
Beilstein Reference: 88199
Merck Reference: 14,4847
MDL No.: MFCD00005651
Formel:
C₁₁H₁₂N₂O₃ Molekulargewicht: 220.23 g/mol Schmelzpunkt: ∼ 298 °C (Zers.) Dichte: 1.405 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00005651 CAS-Nummer: 56-69-9 EINECS: 200-284-8 Merck Index: 13,04873 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...