DL-Cystein, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Amino-3-mercaptopropanoic acid, H-DL-Cys-OH, (±)-Cysteine
M-Clarity™ quality level = MQ100
861677-10G
861677-50G
SIAL861677-10GEA
82
EUR
SIAL861677-10G
SIAL861677-50G
DL-Cystein, Sigma-Aldrich®
DL-Cystein
Formel:
C₃H₇NO₂S Siedepunkt: 254 °C (1013 hPa) Schmelzpunkt: 115 °C Dichte: 1.441 g/cm³ (25 °C) |
MDL Number:
MFCD00004881 CAS-Nummer: 3374-22-9 EINECS: 222-160-2 Merck Index: 12,02850 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...