L(+)-Cystein Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
H-Cys-OH.HCl, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride, L-Cysteine hydrochloride, (R)-(+)-Cysteine hydrochloride
M-Clarity™ Qualitätsstufe = MQ200
C6852-100G
C6852-1KG
C6852-25G
SIALC6852-100GEA
138
EUR
SIALC6852-100G
SIALC6852-1KG
SIALC6852-25G
L(+)-Cystein Hydrochlorid, Sigma-Aldrich®
L(+)-Cystein Hydrochlorid
Formel:
HSCH₂CH(NH₂)COOH·HCl Molekulargewicht: 157.62 g/mol Schmelzpunkt: 175 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00064553 CAS-Nummer: 52-89-1 EINECS: 200-157-7 Merck Index: 13,02810 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...